Epirubicin hydrochloride

Supplier: BioVision

Synonyms: (8S,10S)-10-(((2R,4S,5R,6S)-4-Amino-5-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-6,8,11-trihydroxy-8-(2-hydroxyacetyl)-1-methoxy-7,8,9,10-tetrahydrotetracene-5,12-dione hydrochloride

AB142100-5MG AB142100-25MG
10188-048EA 268.14 USD
10188-048 10188-050
Epirubicin hydrochloride
Epirubicin hydrochloride
Formula: C₂₇H₂₉NO₁₁·HCl
MW: 579.98 g/mol
MDL Number: MFCD00941448
CAS Number: 56390-09-1

Order Now

Specification Test Results

CAS 56390-09-1
Purity ≥95% by HPLC
Form Orange-red powder

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR