Nilotinib
Synonyms:
4-Methyl-N-(3-(4-methyl-1H-imidazol-1-yl)-5-(trifluoromethyl)phenyl)-3-((4-(pyridin-3-yl)pyrimidin-2-yl)amino)benzamide
1750-25
1750-100
10188-036EA
93.39
USD
10188-036
10188-038
Nilotinib
Nilotinib
Formula:
C₂₈H₂₂F₃N₇O MW: 529.5 g/mol |
MDL Number:
MFCD09833716 CAS Number: 641571-10-0 |
Specification Test Results
CAS | 641571-10-0 |
Purity | ≥98% by HPLC |
Form | Off-white solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...