7-Amino-4-(trifluoromethyl)coumarin
Supplier: BioVision
Synonyms:
7-Amino-4-(trifluoromethyl)-2H-chromen-2-one
AB145587-100MG
10191-616EA
160.63
USD
10191-616
7-Amino-4-(trifluoromethyl)coumarin
7-Amino-4-(trifluoromethyl)coumarin
Formula:
C₁₀H₆F₃NO₂ MW: 229.16 g/mol |
MDL Number:
MFCD00006858 CAS Number: 53518-15-3 |
Specification Test Results
CAS | 53518-15-3 |
Purity |
|
Form | Yellow powder |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...