2-Morpholino-8-phenylchromone , EZSolution™

Synonyms: 2-Morpholino-8-phenyl-4-oxo-4H-1-benzopyranLY 2940022-Morpholino-8-phenyl-4H-chromen-4-one

1747-5
10188-028EA 111.89 USD
10188-028
2-Morpholino-8-phenylchromone , EZSolution™
2-Morpholino-8-phenylchromone
Formula: C₁₉H₁₇NO₃
MW: 307.35 g/mol
MDL Number: MFCD00270881
CAS Number: 154447-36-6

Order Now

Specification Test Results

CAS 154447-36-6
Purity ≥98% by HPLC
Form Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR