Rapamycin ≥98% (by HPLC), EZSolution™

Synonyms: (3S,6R,7E,9R,10R,12R,14S,15E,17E,19E,21S,23S,26R,27R,34aS)-9,10,12,13,14,21,22,23,24,25,26,27,32,33,34,34a-Hexadecahydro-9,27-dihydroxy-3-[(1R)-2-[(1S,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethyl]-10,21-dimethoxy-6,8,12,14,20,26-hexamethyl-23,27-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclohentriacontine-1,5,11,28,29(4H,6H,31H)-pentone

1746-5
10188-026EA 254.66 USD
10188-026
Rapamycin ≥98% (by HPLC), EZSolution™
Rapamycin

Liquid

Formula: C₅₁H₇₉NO₁₃
MW: 914.19 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00867594
CAS Number: 53123-88-9
https://us.vwr.com/stibo/web/std.lang.all/06/23/16430623.jpg

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR