Staurosporine , EZSolution™

Synonyms: Antibiotic AM-2282

1745-01
10188-024EA 111.84 USD
10188-024
Staurosporine , EZSolution™
Staurosporine
Formula: C₂₈H₂₆N₄O₃
MW: 466.541 g/mol
MDL Number: MFCD18252446
CAS Number: 62996-74-1

Order Now

Specification Test Results

CAS 62996-74-1
Purity ≥98% by HPLC
Form Liquid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR