GW2580

Synonyms: 5-[3-Methoxy-4-[(4-methoxybenzyl)oxy]benzyl]pyrimidine-2,4-diamine5-(3-Methoxy-4-((4-methoxybenzyl)oxy)benzyl)pyrimidine-2,4-diamine

1740-25 1740-5
10188-012EA 361.93 USD
10188-012 10191-664
GW2580
GW2580
Formula: C₂₀H₂₂N₄O₃
MW: 366.42 g/mol
MDL Number: MFCD12024701
CAS Number: 870483-87-7

Order Now

Specification Test Results

CAS 870483-87-7
Purity ≥99% by HPLC
Form Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR