Oridonin

Supplier: BioVision

Synonyms: (1α,6β,7α,14R)-1,5,6,14-Tetrahydroxy-4,4-dimethyl-8-methylenedecahydro-1H-6,11b-(epoxymethano)-6a,9-methanocyclohepta[a]naphthalen-7(8H)-oneIsodonol

AB142094-5MG AB142094-25MG
10187-984EA 191.54 USD
10187-984 10187-982
Oridonin
Oridonin
Formula: C₂₀H₂₈O₆
MW: 364.4 g/mol
MDL Number: MFCD00221762
CAS Number: 28957-04-2

Order Now

Specification Test Results

CAS 28957-04-2
Purity ≥97% by HPLC
Form Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR