PI103

Synonyms: 3-(4-Morpholinopyrido[3',2':4,5]furo[3,2-d]pyrimidin-2-yl)phenol

1728-5 1728-1
10187-976EA 396.95 USD
10187-976 10187-974
PI103
PI103
Formula: C₁₉H₁₆N₄O₃
MW: 348.4 g/mol
MDL Number: MFCD11983145
CAS Number: 371935-74-9

Order Now

Specification Test Results

CAS 371935-74-9
Purity ≥98% by HPLC
Form Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR