Tubastatin A

Supplier: BioVision

Synonyms: N-Hydroxy-4-((2-methyl-3,4-dihydro-1H-pyrido[4,3-b]indol-5(2H)-yl)methyl)benzamide 2,2,2-trifluoroacetate

AB141415-5MG AB141415-1MG
10187-966EA 1938.15 USD
10187-966 10187-964
Tubastatin A
Tubastatin A
Formula: C₂₂H₂₂F₃N₃O₄
MW: 449.4230 g/mol
MDL Number: MFCD24873137
CAS Number: 1239262-52-2

Order Now

Specification Test Results

CAS 1239262-52-2
Purity ≥98% by HPLC
Form Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR