3-Isobutyl-1-methylxanthine

Synonyms: 2,6-Dihydroxy-3-isobutyl-1-methylpurine3,7-Dihydro-3-isobutyl-1-methyl-1H-purine-2,6-dione3-Isobutyl-1-methyl-3,7-dihydro-1H-purine-2,6-dione

1714-100
10187-952EA 75.89 USD
10187-952
3-Isobutyl-1-methylxanthine
3-Isobutyl-1-methylxanthine
Formula: C₁₀H₁₄N₄O₂
MW: 222.25 g/mol
MDL Number: MFCD00005584
CAS Number: 28822-58-4

Order Now

Specification Test Results

CAS 28822-58-4
Purity ≥98% by HPLC
Form White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR