Akt inhibitor VIII Isozyme-Selective

Synonyms: Akti-1/21,3-Dihydro-1-[1-[4-(6-phenyl-1H-imidazo[4,5-g]quinoxalin-7-yl)phenylmethyl]-4-piperidinyl]-2H-benzimidazol-2-one

1708-1
10187-938EA 163.2 USD
10187-938
Akt inhibitor VIII Isozyme-Selective
Akt inhibitor VIII
Formula: C₃₄H₂₉N₇O
MW: 551.65 g/mol
MDL Number: MFCD09026909
CAS Number: 612847-09-3

Order Now

Specification Test Results

CAS 612847-09-3
Purity ≥97% by HPLC
Form Light yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR