BML-277 Chk2 inhibitor
Supplier: BioVision
Synonyms:
2-(4-(4-Chlorophenoxy)phenyl)-1H-benzo[d]imidazole-6-carboxamide
, 2-(4-(4-Chlorophenoxy)phenyl)-1H-benzimidazole-5-carboxamide
, Chk2 Inhibitor II
AB142086-1MG
10187-924EA
298.31
USD
10187-924
BML-277 Chk2 inhibitor
BML-277 Chk2 inhibitor
Formula:
C₂₀H₁₄ClN₃O₂ MW: 363.8 g/mol |
MDL Number:
MFCD08276917 CAS Number: 516480-79-8 |
Specification Test Results
CAS | 516480-79-8 |
Purity | ≥98% by HPLC |
Form | Pale yellow solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...