Resiquimod

Synonyms: R-8481-(4-Amino-2-(ethoxymethyl)-1H-imidazo[4,5-c]quinolin-1-yl)-2-methylpropan-2-ol4-Amino-2-(ethoxymethyl)-a,a-dimethyl-1H-imidazo[4,5-c]quinoline-1-ethanol1-[4-Amino-2-(ethoxymethyl)-1H-imidazo[4,5-c]quinolin-1-yl]-2-methylpropan-2-ol

1699-5 1699-25
10187-920EA 251 USD
10187-920 10187-918
Resiquimod
Resiquimod
Formula: C₁₇H₂₂N₄O₂
MW: 314.389 g/mol
MDL Number: MFCD00937759
CAS Number: 144875-48-9

Order Now

Specification Test Results

CAS 144875-48-9
Purity ≥98% by HPLC
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR