Mevastatin
Synonyms:
(1S,7S,8S,8aR)-8-(2-((2R,4R)-4-Hydroxy-6-oxotetrahydro-2H-pyran-2-yl)ethyl)-7-methyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl (S)-2-methylbutanoate
, Compactin
1694-250
1694-50
10187-906EA
81.6
USD
10187-906
10187-904
Mevastatin
Mevastatin
Formula:
C₂₃H₃₄O₅ MW: 390.52 g/mol |
MDL Number:
MFCD05662341 CAS Number: 73573-88-3 |
Specification Test Results
CAS | 73573-88-3 |
Purity | ≥95% by HPLC |
Form | White solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...