1,1-Dimethylbiguanide hydrochloride

Synonyms: AMPK Signaling Activator VIIMetformin1,1-Dimethylbiguanide HydrochlorideMetformin hydrochloride

1691-5G
10191-658EA 130.25 USD
10191-658
1,1-Dimethylbiguanide hydrochloride
1,1-Dimethylbiguanide hydrochloride
Formula: C₄H₁₂ClN₅
MW: 165.625 g/mol
MDL Number: MFCD00012582
CAS Number: 1115-70-4

Order Now

Specification Test Results

CAS 1115-70-4
Purity ≥99%
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR