QS11

Supplier: BioVision

AB141408-5MG
10187-844EA 436.53 USD
10187-844
QS11
QS11
Formula: C₃₅H₃₃N₅O₂
MW: 567.7 g/mol
MDL Number: MFCD18086864
CAS Number: 944328-88-5

Order Now

Specification Test Results

CAS 944328-88-5
Purity ≥98% by HPLC
Form White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR