(2'Z,3'E)-6-Bromoindirubin-3'-oxime

Synonyms: 6-Bromoindirubin-3'-oximeBIO(2Z,3E)-6'-Bromo-3-(hydroxyimino)-[2,3'-biindolinylidene]-2'-one6BIO

1673-1
10187-834EA 116.75 USD
10187-834
(2'Z,3'E)-6-Bromoindirubin-3'-oxime
(2'Z,3'E)-6-Bromoindirubin-3'-oxime
Formula: C₁₆H₁₀BrN₃O₂
MW: 356.18 g/mol
MDL Number: MFCD08705318
CAS Number: 667463-62-9

Order Now

Specification Test Results

CAS 667463-62-9
Purity ≥98%
Form Crystalline solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR