2-Morpholino-8-phenylchromone

Synonyms: 2-Morpholino-8-phenyl-4-oxo-4H-1-benzopyranLY 2940022-Morpholino-8-phenyl-4H-chromen-4-one

1667-25 1667-5
10187-820EA 99.08 USD
10187-820 10187-818
2-Morpholino-8-phenylchromone
2-Morpholino-8-phenylchromone
Formula: C₁₉H₁₇NO₃
MW: 307.35 g/mol
MDL Number: MFCD00270881
CAS Number: 154447-36-6

Order Now

Specification Test Results

CAS 154447-36-6
Purity ≥98% by HPLC
Form Off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR