2-Morpholino-4H-pyrimido[2,1-a]isoquinolin-4-one

Synonyms: Compound 401

1657-5
10187-808EA 201.98 USD
10187-808
2-Morpholino-4H-pyrimido[2,1-a]isoquinolin-4-one
2-Morpholino-4H-pyrimido[2,1-a]isoquinolin-4-one
Formula: C₁₆H₁₅N₃O₂
MW: 281.32 g/mol
MDL Number: MFCD10568147
CAS Number: 168425-64-7

Order Now

Specification Test Results

CAS 168425-64-7
Purity ≥97% by HPLC
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR