TWS119

Supplier: BioVision

Synonyms: 3-[[6-(3-Aminophenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-yl]oxy]phenol3-((6-(3-Aminophenyl)-7H-pyrrolo[2,3-d]pyrimidin-4-yl)oxy)phenol

AB142075-2MG
10187-804EA 341.18 USD
10187-804
TWS119
TWS119
Formula: C₁₈H₁₄N₄O₂
MW: 318.34 g/mol
MDL Number: MFCD09037535
CAS Number: 601514-19-6

Order Now

Specification Test Results

CAS 601514-19-6
Purity ≥95% by HPLC
Form Slightly pink solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR