Procaspase-3-activator (PAC-1)

Supplier: BioVision

Synonyms: 4-(Phenylmethyl)[[2-hydroxy-3-(2-propenyl)phenyl]methylene]hydrazide-1-piperazine acetic acid(4-benzylpiperazino)acetic acid-(3-allyl-2-hydroxybenzylidene)hydrazide

AB142074-10
10187-802EA 256.59 USD
10187-802
Procaspase-3-activator (PAC-1)
Procaspase-3 Activator (PAC1, (4-Benzylpiperazino)acetic acid-(3-allyl-2-hydroxybenzylidene)hydrazide)

Order Now

Specification Test Results

CAS 315183-21-2
Purity ≥95% by HPLC
Form White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR