AGK2

Supplier: BioVision

Synonyms: 2-Cyano-3-(5-(2,5-dichlorophenyl)furan-2-yl)-N-(quinolin-5-yl)acrylamide2-Cyano-3-[5-(2,5-dichlorophenyl)furan-2-yl]-N-(quinolin-5-yl)acrylamide

AB142073-10MG
10187-796EA 367.72 USD
10187-796
AGK2
AGK2
Formula: C₂₃H₁₃Cl₂N₃O₂
MW: 434.28 g/mol
MDL Number: MFCD01909444
CAS Number: 304896-28-4

Order Now

Specification Test Results

CAS 304896-28-4
Purity ≥95% by HPLC
Form Yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR