AEBSF hydrochloride (4-(2-Aminoethyl)benzenesulfonylfluoride hydrochloride)

Supplier: BioVision

Synonyms: 4-(2-Aminoethyl)benzenesulfonyl fluoride hydrochloride4-(2-Aminoethyl)benzenesulphonyl fluoride hydrochloride

AB141403-200MG 1644-1000
10187-768EA 618.77 USD
10187-768 10187-770
AEBSF hydrochloride (4-(2-Aminoethyl)benzenesulfonylfluoride hydrochloride)
AEBSF hydrochloride (4-(2-Aminoethyl)benzenesulphonylfluoride hydrochloride)
Formula: C₈H₁₁ClFNO₂S
MW: 239.69 g/mol
MDL Number: MFCD00132962
CAS Number: 30827-99-7

Order Now

Specification Test Results

CAS 30827-99-7
Purity ≥97% by HPLC
Form White solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR