Lapatinib ditosylate

Synonyms: GW57216 ditosylate monohydrateTykerb ditosylateGW216 ditosylate monohydrateN-(3-Chloro-4-((3-fluorobenzyl)oxy)phenyl)-6-(5-(((2-(methylsulphonyl)ethyl)amino)methyl)furan-2-yl)quinazolin-4-amine bis(4-methylbenzenesulphonate) hydrateN-(3-Chloro-4-((3-fluorobenzyl)oxy)phenyl)-6-(5-(((2-(methylsulfonyl)ethyl)amino)methyl)furan-2-yl)quinazolin-4-amine bis(4-methylbenzenesulfonate) hydrate

1624-100 1624-25
10187-752EA 169.02 USD
10187-752 10187-750
Lapatinib ditosylate
Lapatinib ditosylate
Formula: C₂₉H₂₆ClFN₄O₄S·₂C₇H₈O₃S
MW: 925.46 g/mol (anhy)
MDL Number: MFCD09264195
CAS Number: 388082-78-8

Order Now

Specification Test Results

CAS 388082-78-8
Purity ≥99% by HPLC
Form Yellow solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR