Canertinib dihydrochloride ≥99%

Supplier: BioVision

Synonyms: CI-1033N-[4-[(3-Chloro-4-fluorophenyl)amino]-7-[3-(4-morpholinyl)propoxy]-6-quinazolinyl]-2-propenamide dihydrochloridePD-183805

AB141402-50MG AB141402-5MG
10187-740EA 789.07 USD
10187-740 10187-738
Canertinib dihydrochloride ≥99%
Canertinib dihydrochloride

Canertinib is an irreversible tyrosine kinase inhibitor. It not only inhibits tyrosine phosphorylation but also enhances ubiquitinylation and accelerates endocytosis and subsequent intracellular destruction of ErbB-2 molecules. Canertinib alkylates a cysteine residue specific to ErbB receptors. The degradative pathway of ErbB receptor tyrosine kinases stimulated by tyrosine kinase inhibitors appears to be chaperone mediated, and thus is similar to the pathways activated by the heat shock protein 90 (Hsp90) antagonist geldanamycin and by stress-induced mechanisms.

Formula: C₂₄H₂₅ClFN₅O₃·2HCl
MW: 558.86 g/mol
Storage Temperature: Freezer
MDL Number: MFCD09954112
CAS Number: 289499-45-2

Order Now

Specification Test Results

CAS 289499-45-2
Purity ≥99%
Form Off-white to pale green solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR