VX-702

Supplier: BioVision

Synonyms: 6-[(Aminocarbonyl)(2,6-difluorophenyl)amino]-2-(2,4-difluorophenyl)-3-pyridinecarboxamide

AB142057-1MG AB142057-5MG
10187-702EA 2305.93 USD
10187-702 10187-700
VX-702
VX-702
Formula: C₁₉H₁₂F₄N₄O₂
MW: 404.32 g/mol
MDL Number: MFCD11616590
CAS Number: 479543-46-9

Order Now

Specification Test Results

CAS 745833-23-2
Purity ≥99% by HPLC
Form White soild

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR