Deforolimus
1587-25
1587-5
10187-658EA
1097.45
USD
10187-658
10187-660
Deforolimus
Deforolimus
CAS Number:
572924-54-0 |
Specification Test Results
CAS | 572924-54-0 |
Purity | ≥99% by HPLC |
Form | White to off-white solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...