AZD1152-HQPA

Supplier: BioVision

Synonyms: 2-(5-((7-(3-(Ethyl(2-hydroxyethyl)amino)propoxy)quinazolin-4-yl)amino)-1H-pyrazol-3-yl)-N-(3-fluorophenyl)acetamideBarasertib

AB142049-25MG AB142049-5MG
10187-642EA 528.57 USD
10187-642 10187-640
AZD1152-HQPA
AZD1152-HQPA
Formula: C₂₆H₃₀FN₇O₃
MW: 507,56 g/mol
MDL Number: MFCD17392583
CAS Number: 722544-51-6

Order Now

Specification Test Results

CAS 722544-51-6
Purity ≥99% by HPLC
Form Crystalline solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR