Tyrphostin AG 1295
1571-5
10187-600EA
239.32
USD
10187-600
Tyrphostin AG 1295
Tyrphostin AG 1295
CAS Number:
71897-07-9 |
Specification Test Results
CAS | 71897-07-9 |
Purity | ≥99% |
Form | Yellow solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...