Rapamycin ≥99%
Synonyms:
(3S,6R,7E,9R,10R,12R,14S,15E,17E,19E,21S,23S,26R,27R,34aS)-9,10,12,13,14,21,22,23,24,25,26,27,32,33,34,34a-Hexadecahydro-9,27-dihydroxy-3-[(1R)-2-[(1S,3R,4R)-4-hydroxy-3-methoxycyclohexyl]-1-methylethyl]-10,21-dimethoxy-6,8,12,14,20,26-hexamethyl-23,27-epoxy-3H-pyrido[2,1-c][1,4]oxaazacyclohentriacontine-1,5,11,28,29(4H,6H,31H)-pentone
An immunosuppressant which selectively blocks signaling that leads to p70 S6 kinase activation (IC₅₀ = 50 pM).
- White to Off-white solid
- Soluble in DMSO and EtOH
Inhibits lymphokine-induced cell proliferation at the G1 phase and induces apoptosis in a murine B cell line.
Formula:
C₅₁H₇₉NO₁₃ MW: 914.19 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD00867594 CAS Number: 53123-88-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...