Mifepristone

Synonyms: RU 486

1561-50
10187-550EA 244 USD
10187-550
Mifepristone
Mifepristone
Formula: C₂₉H₃₅NO₂
MW: 429.60 g/mol
CAS Number: 84371-65-3

Order Now

Specification Test Results

CAS 84371-65-3
Purity ≥99% by TLC
Form Yellow soild

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR