Rosiglitazone

Synonyms: 5-(4-(2-(Methyl(pyridin-2-yl)amino)ethoxy)benzyl)thiazolidine-2,4-dione

1559-100 1559-50 1559-5
10187-544EA 344.42 USD
10187-544 10187-542 10187-540
Rosiglitazone
Rosiglitazone
Formula: C₁₈H₁₉N₃O₃S
MW: 357.43 g/mol
MDL Number: MFCD00871760
CAS Number: 122320-73-4

Order Now

Specification Test Results

CAS 122320-73-4
Purity ≥99%
Form White to off-white solid

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR