Ethacrynic acid
1556-1000
10187-530EA
99.08
USD
10187-530
Ethacrynic acid
Ethacrynic acid
GST Inhibitor-2
Formula:
C₁₃H₁₂Cl₂O₄ |
MDL Number:
MFCD00056693 CAS Number: 58-54-8 |
Specification Test Results
CAS | 58-54-8 |
Purity | ≥99% |
Form | White solid |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...