Mouse Recombinant VEGF165 (from E. coli)
Supplier: BioVision
AB285568-10UG AB285568-50UG AB285568-1MG
10190-416EA 589.89 USD
10190-416 10190-418 10190-420
Mouse Recombinant VEGF165 (from E. coli)
Proteins and Peptides
>98% Pure active Recombinant murine VEGF165.
Determined by the mitogenic activity on human μmbilical vein endothelial cells and bovine aortic endothelial cells. Mitogenic activity was obtained for stimulation of 3H-thymidine incorporation and cell proliferation in a concentration range of 1.0 to 5.0 ng/ml.
VEGF (Vascular Endothelial Growth Factor) is a homodimeric protein secreted by a variety of Vascularized tissues. VEGF stimulates endothelial cell growth, angiogenesis, and capillary permeability. Murine VEGF is a 39 kDa homodimeric protein consisting of two 165 amino acid polypeptide chains.