Human Recombinant TWEAK (from E. coli)
Supplier: BioVision
4360-10 4360-1000
10190-406EA 244 USD
10190-406 10190-408
Human Recombinant TWEAK (from E. coli)
>98% Pure active Recombinant human TWEAK.
The ED₅₀ as determined by the dose-dependent stimulation of IL-8 production by human PBMC is less than 10 ng/ml.
Human TWEAK is a new secreted ligand in the tumor necrosis factor family that weakly induces apoptosis. TWEAK, like many other members of the TNF family, is able to induce interleukin-8 synthesis in a number of cell lines. The recombinant human TWEAK is a 17.0 kDa protein, comprising the TNF-like extraCellular domain of TWEAK