Heating Bath Circulators, CC-Range, Huber
Supplier: Huber
2049.0002.01 2060.0002.01 2053.0002.01 2005.0002.01 2051.0002.01 2037.0058.01 2004.0002.01 2056.0002.01 2003.0002.01 2007.0001.01 2006.0003.01 2058.0002.01 2059.0002.01 2052.0002.01 2050.0002.01 2057.0002.01
10220-990EA 6147.22 USD
10220-990 10221-706 10221-914 10220-982 10220-988 10220-976 10220-998 10220-978 10220-830 10220-994 10221-002 10221-012 10221-010 10220-828 10221-006 10221-910
Heating Bath Circulators, CC-Range, Huber
Circulating Thermostats Closed Circulator Systems Heating Circulators
Heating circulation thermostats are specially designed for temperature control of externally attached applications with a small volume
The new multi-touch controller Pilot ONE offers a brilliant 5.7" touchscreen with graphic functions and a comfortable navigation menu in 11 languages. All important operating parameters and temperature values are neatly displayed on the touchscreen. Thanks to the new favourites menu, the one-click operation and the integrated technical glossary the operation is very easy, just like on a smartphone. Integrated USB and Ethernet ports allows connection to a PC or network, e.g. for remote control or data transmission.
CC-range has the following advantages and functions: Pilot ONE controller with Plug & Play technology, precise temperature control, 5.7“ touchscreen and comfortable menu navigation, overtemperature and low level protection, safety class III/FL (DIN 12876).
The units are fitted with bath tanks made of transparent polycarbonate. The thermostats have M16x1 pump connections at the rear side as well as a stainless steel bath cover with a filling opening. All models are fitted with an overtemperature and low level protection system according to safety class III/FL (DIN 12876) for use with flammable fluids. A powerful circulation pump ensures optimal heat transfer to external attached applications.
Ordering information: Depending on the model, heating circulation thermostats could be supplied with a mini-USB cable, adapter nom. dia 12 mm, bath cover, dummy plugs, sleeve nuts thread M16x1, connection tube silicon, power supply cable. Please contact your VWR representative for standard delivery specification.