Immersion Circulators, CC-Range, Huber
Supplier: Huber
2000.0024.01 2061.0002.01
10220-966EA 4349.36 USD
10220-966 10220-968
Immersion Circulators, CC-Range, Huber
Circulating Thermostats Open Circulator Systems Thermostat Controllers
Immersion circulators are suitable for controlling the temperature of any bath as the screw clamp allows the thermostats to be easily mounted on different vessels
The new multi-touch controller Pilot ONE offers a brilliant 5.7" touchscreen with graphic functions and a comfortable navigation menu in 11 languages. All important operating parameters and temperature values are neatly displayed on the touchscreen. Thanks to the new favourites menu, the one-click operation and the integrated technical glossary the operation is very easy, just like on a smartphone. Integrated USB and Ethernet ports allows connection to a PC or network, e.g. for remote control or data transmission.
CC-range has the following advantages and functions: Pilot ONE controller with Plug and Play technology, precise temperature control, 5.7“ touchscreen and comfortable menu navigation, overtemperature and low level protection, safety class III/FL (DIN 12876)
Minus temperatures are achieved in combination with a refrigerated bath or immersion cooler. All models are fitted with a powerful pressure/suction pump and meet the requirements of Safety Class III (FL) for use with flammable fluids. A pump adapter for external temperature control and cooling coils for a cooling water connection are available as accessories.
Ordering information: Depending on the model, immersion thermostat could be supplied with a mini-USB cable, screw clamp. Please contact your VWR representative for standard delivery specification.