Would you like to log in again?
Grignard Synthesis of Benzoic Acid
Supplier: ALDON CORP SE
IS8022
Prepare a carboxylic acid by the Grignard method.
  • Classic AP Chemistry Experiment
  • Instructions Included
Once the Grignard reagent is prepared, it can be used to prepare the target compound. In this experiment the target compound is a carboxylic acid. To prepare a carboxylic acid, the Grignard reagent is carboxylated in a reaction with dry ice. After the reaction is complete, hot water is added to evaporate any remaining solvent. Acid is added to dissolve the magnesium salts and liberate the carboxylic acid. Once the solution is cooled, the acid can be collected. In this experiment, we will prepare a carboxylic acid (benzoic acid) by the Grignard method.
Ordering information: Includes Magnesium Metal Turnings, Tetrahydrofuran, Bromobenzene, Hydrochloric Acid, and Decolorizing Carbon.
ORDER
Expand All / Collapse All
Description Includes No. of Lab Groups Supplier No. VWR Catalog Number Unit Price Quantity
Grignard Synthesis of Benzoic Acid Teacher's Guide and Student Copymaster 15 IS8022 470163-110 Kit of 1 Retrieving
Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page
Q and A
Product Questions & Answers
Sort by: Most Helpful | Newest | Oldest
Items Per Page: 5  10  20 
2  3  4  Next
Can't find an answer to your question?  Ask us a question
Ask us a question:
Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question
Q&A Guidelines & Tips
If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.
When writing your question or answer, please follow these guidelines:
  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English
All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s
We reserve the right not to post your question if it contains any of the following
  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details
Select the article for which you are looking for Certificate
Indicate the Lot Number. This field is mandatory.
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty