Keep my session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-KNGP Mouse Monoclonal Antibody [clone: 24F9]

Supplier: Young In Frontier
LF-MA0204
Kininogens are precursors for kinin, and the two main types of them are high-molecular weight kininogen (HMWK) and low-molecular weight kininogen (LMWK). HMWK also known as the Williams-Fitzgerald-Flaujeac factor is a protein from the blood coagulation system as well as the kinin-kallikrein system. It acts mainly as a cofactor on coagulation and inflammation, and has no intrinsic catalytic activity. LMWK is produced locally by numerous tissues, and secreted together with tissue kallikrein.
Kininogens are synthesized in the liver and circulate in the plasma and other body fluids.
The kinins are pharmacologically active polypeptides released in the tissues and body fluids as a result of the enzymatic action of kallikreins on kininogens. The kinin family includes bradykinin (BK) (Arg-Pro-Pro-gly-Phe-Ser-Pro-Phe-Arg), kallidin (Lys-Arg-Pro-Pro-Gly-Phe-Ser-Pro-Phe-Arg) and methionyl-lysyl-BK (Met-Lys-Arg-Pro-Pro-Gly-Phe-Arg). Kallidin and methionyl-lysyl-BK are converted into BK by aminopeptidases present in plasma and urine. Active tissue kallikrein acts on LMWK to release kallidin. The plasma kallikrein is found in circulation in an inactive form, which is known as prekallikrein or Fletcher factor.
BK and kallidin act through activation of G-protein-coupled constitutive B(2) or inducible kinin B(1) receptors linked to signaling pathways involving increased intracellular Ca concentrations and/or release of mediators including arachidonic acid metabolites, NO and EDHF (Endothelium-derived hyperpolarizing factor). In the cardiovascular system, the kallikrein-kinin system exerts a fine control of vascular smooth muscle tone and arterial blood pressure, and plays a significant cardioprotective effect.

Western Blot: 0.5 μg/ml;

Type: Primary
Antigen: KNGP
Clonality: Monoclonal
Clone: 24F9
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human




 

ORDER

Expand All / Collapse All
Size Supplier No. VWR Catalog Number Unit Price Quantity
100 µL LF-MA0204 10219-228 Each (100µl) Retrieving Restricted

Temperature sensitive This Item is temperature-controlled and may have specific temperature and storage requirements for shipping/delivery.

restricted In order to process your orders without delay, we request that you provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide identification that includes your business name and shipping address via email CMD_NA@vwr.com or fax 484.881.5997 referencing your VWR account number . Acceptable forms of identification are:

  • State issued document with your organization's Federal Tax ID Number
  • State issued document with your organization's Resale Tax ID Number
  • City or County issued Business License
  • State Department of Health Services License
  • Any other ID issued by the State that includes the business name and address

* Please note if your account is within the State of California two of these pieces of identification will be required.
VWR will not lift restrictions for residential shipping addresses.

Generate Barcodes Barcode Label Format: 
Generate PDF Catalog Page

SPECIFICATIONS

Antigen Symbol KNGP
Antigen Name Kininogen Precursor
Reactivity Human
Conjugation Unconjugated
Clonality Monoclonal
Antibody Type Primary
Clone 24F9
Host Mouse
Isotype IgG
Western Blot Yes
ELISA Yes
Concentration 1 mg/ml
Cross Adsorption none
Immunogen Protein purified from Human plasma
Purification Protein G purified
Storage Temperature Store for 1 year at -20°C from date of shipment
Shipping Temperature Ice

Q and A

Product Questions & Answers

Sort by: Most Helpful | Newest | Oldest

Items Per Page: 5  10  20 
2  3  4  Next

Can't find an answer to your question?  Ask us a question

Ask us a question:

Please enter Public Name.
(Not published) The email address you have provided is invalid. Please enter a valid email address.
Please enter Question

Q&A Guidelines & Tips

If you would like to share feedback with us about pricing, delivery or other customer service issues, please contact customer service directly.

When writing your question or answer, please follow these guidelines:

  • Do: Make sure your question is directly related to the product
  • Do: Be specific and provide as many details as possible
  • Do: Include t he model number (if applicable)
  • Do: Write your question in English

All submitted questions and answers are subject to the terms set forth in our Terms and Condition.s

We reserve the right not to post your question if it contains any of the following

  • Any information not related to the actual merchandise or topic
  • Obscenities, discriminatory language or other language not suitable for a public forum
  • Advertisements or content
  • Email addresses, physical addresses or other forms of contact information
  • Critical or spiteful comments on other questions or answers posted on the page or their authors
  • Anything that VWR deems to be inappropriate, at VWR discretion
Enter Details

Select the article for which you are looking for Certificate

Indicate the Lot Number. This field is mandatory.

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty