Anti-SELM Mouse Monoclonal Antibody [clone: 10C1]

Supplier: Young In Frontier
LF-MA0058
10219-050EA 413.6 USD
10219-050
Anti-SELM Mouse Monoclonal Antibody [clone: 10C1]
A growing body of evidence relates selenium to cancer prevention, immune system function, male fertility, cardiovascular disorder, control of the aging and neurodiseases process. Selenoproteins are thought to be responsible for the majority of these biomedical effects of selenium. Approximately 17 selenoproteins have been identified until now. Although the function of many selenoproteins are unknown, some play important roles in anti-oxidant mechanisms. It has been also implicated in the regulation of  signaling pathways through catalysis of thiol/ disulfide exchange.
Selenoprotein M(SelM) is especially expressed in a mammalian brain and is localized to the perinuclear structures (Golgi/ER). The roles of selM have not been clearly identified until present time.

Western Blot: 1:2000; Immunoprecipitation: 1-2 ?l;

Type: Primary
Antigen: SELM
Clonality: Monoclonal
Clone: 10C1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR