RX-100™ Bifocal Safety Glasses, Elvex®
Supplier: Elvex
RX-100-3.0 RX-100-1.5
33200-108CS 791.78 USD
33200-108 33200-102
RX-100™ Bifocal Safety Glasses, Elvex®
Safety Glasses
Safety glasses are made of hard coated polycarbonate lenses, clear or gray.
  • Temples adjust in four different lengths
  • Temples have panascopic angle adjustment for perfect fit
  • Lenses have integrated side shields
Feature-packed bifocal with a unitary lens and provides wide, unobstructed field of vision.
Certifications: Eyewear meets ANSI Z87 and CE-EN-166 standards.