Anti-CHEK2 Rabbit Polyclonal Antibody

Supplier: Abnova
PAB25431
10674-450EA 426.15 USD
10674-450
Anti-CHEK2 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against synthetic phosphopeptide of CHEK2.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: CHEK2
Clonality: Polyclonal
Clone: 0
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity:
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR