Anti-DACT1 Mouse Monoclonal Antibody [clone: 5D8]

Supplier: Abnova
H00051339-M03
10583-438EA 335 USD
10583-438
Anti-DACT1 Mouse Monoclonal Antibody [clone: 5D8]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant DACT1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: DACT1
Clonality: Monoclonal
Clone: 5D8
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR