Anti-IL17C Mouse Polyclonal Antibody

Supplier: Abnova
H00027189-B01
10561-608EA 335 USD
10561-608
Anti-IL17C Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antibody raised against a full-length human IL17C protein.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: IL17C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR