Anti-HSPB7 Mouse Monoclonal Antibody [clone: 1G12]

Supplier: Abnova
H00027129-M02
10589-496EA 335 USD
10589-496
Anti-HSPB7 Mouse Monoclonal Antibody [clone: 1G12]
Antibodies
Mouse monoclonal antibody raised against a full-length recombinant HSPB7.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: HSPB7
Clonality: Monoclonal
Clone: 1G12
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR