Anti-SASH1 Mouse Monoclonal Antibody [clone: X1]

Supplier: Abnova
H00023328-M02
10600-664EA 335 USD
10600-664
Anti-SASH1 Mouse Monoclonal Antibody [clone: X1]
Antibodies
Mouse monoclonal antibody raised against a partial recombinant SASH1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: SASH1
Clonality: Monoclonal
Clone: X1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR