Anti-HAVCR1 Mouse Monoclonal Antibody [clone: 1G1]

Supplier: Abnova
H00026762-M04
10588-326EA 335 USD
10588-326
Anti-HAVCR1 Mouse Monoclonal Antibody [clone: 1G1]
Antibodies
Mouse monoclonal antibody raised against a full-length recombinant HAVCR1.

Recommended Dilutions: Western Blot: 1-5 ?g/ml

Type: Primary
Antigen: HAVCR1
Clonality: Unconjugated
Clone: Monoclonal
Conjugation: 1G1
Epitope:
Host: IgG2b kappa
Isotype: Mouse
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR