Anti-PDXDC1 Rabbit Polyclonal Antibody

Supplier: Abnova
H00023042-D01
10591-022EA 376 USD
10591-022
Anti-PDXDC1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody raised against a full-length human PDXDC1 protein.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: PDXDC1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR