Anti-KDM2A Mouse Polyclonal Antibody

Supplier: Abnova
H00022992-B01
10556-388EA 257.43 USD
10556-388
Anti-KDM2A Mouse Polyclonal Antibody
Antibodies
Mouse polyclonal antibody raised against a full-length human FBXL11 protein.

Recommended Dilutions: Western Blot: 1:500-1:1000

Type: Primary
Antigen: KDM2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR